• USA Home
  • Z175889 - Aldrich® Hartzler condenser
EMAIL THIS PAGE TO A FRIEND
Z175889 Aldrich
Aldrich® Hartzler condenser
column H 340 mm, Joint: ST/NS 19/38
Purchase
Properties
Related Categories Coiled High-Capacity Condensers, Glass Condensers, Glassware, High-Capacity Condensers, Labware More...
Joint   Joint: ST/NS19/38
column H   340 mm
Description
General description
Lower primary condenser of bulbous portions connected with each other and with an upper curvilinear column.
Footnote
Designed by Harold E. Hartzler; Goshen, IN.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?