• USA Home
  • Z176079 - Lecture-bottle valve
EMAIL THIS PAGE TO A FRIEND
Z176079 Aldrich
Lecture-bottle valve
steel (carbon-plated)
Purchase
Description
General description
Diffusion resistant, diaphragm seal, packless on/off valve. 3/8 in. NPTM valve thread. CGA 180M inlet/110F outlet connection.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?