• USA Home
  • Z176559 - Round-bottom oven flask
EMAIL THIS PAGE TO A FRIEND
Z176559 Aldrich
Round-bottom oven flask
Purchase
Properties
Related Categories Aldrich, Aldrich Flasks, Aldrich Round-Bottom Flasks, Flasks, Labware,
material   borosilicate glass
  flask round bottom
sterility   non-sterile
packaging   pack of 1
mfr. no.   Sigma-Aldrich
capacity   4000 mL
Description
General description
2- and 4-liter flasks have support tube for use with large-capacity oven.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?