• USA Home
  • Z176672 - Aldrich® cold plate stirrer
EMAIL THIS PAGE TO A FRIEND
Z176672 Aldrich
Aldrich® cold plate stirrer
AC/DC input 115 V, Analog cold plate stirrer
Purchase
Description
General description
These portable, low-temperature control devices utilize efficient, solid-state, thermoelectric modules to control temperatures from +25 to -20 °C. Unique, dual heat exchanger utilizes either fan forced air or tap water for cooling. Connects to water source using 1/4 in. I.D. tubing. Stirring models have a built-in, adjustable-speed, magnetic stirrer to keep solutions at equilibrium. Useful for reaction kinetics, recrystallizations, cold traps, freezing point determinations, controlling exothermic reactions, condenser for distillations, freezing biological and blood specimens, cold bath or storage chambers. Not CE compliant.
Specifications:
• Rapid cooling to -10 °C (forced air) or -20 °C (tap water), 130 BTU/h initial rate
• Cooling to -40 °C possible by circulating coolant
• Equilibrium reached in 2 to 3 min
• 3 in. diam. aluminum top plate handles solutions up to 200 mL
• 18 gauge aluminum cabinet, 7 W × 5 D × 5 in. H
• Wt = 8 lb
Analog models have adjustable power supply to control cooling.
Digital models have dial-in temperature control with digital setpoint in 1 °C increments, automatic thermistor temperature control (order thermistor sensors separately below), and digital meter readout to 0.1 °C with 0.1 °C accuracy.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?