• USA Home
  • Z179183 - AMPCO® Non-sparking safety tools
EMAIL THIS PAGE TO A FRIEND
Z179183 Aldrich
AMPCO® Non-sparking safety tools
pieces size 6
Purchase
Properties
Related Categories AMPCO, AMPCO Non-Sparking Safety Tools, AMPCO Tools, Labware, Manufacturer Browser,
mfr. no.   AMPCO M47
pieces size   6
Description
General description
Nonsparking, nonmagnetic, corrosion-resistant tools produced from nonferrous alloys. FM approved. Satisfies OSHA guidelines suggesting that nonsparking tools be used in locations where flammable vapors and combustible residues are present. Certain chemicals, such as acetylene, amatol, and similar liquid gases, may react with copper alloys to form metalloids that negate the nonsparking properties of these alloys. The 2, 3, 4, and 6 piece sets come in a vinyl case with nonjamming nonmetallic zipper. The 16 piece set comes in a plastic box.
Legal Information
AMPCO is a registered trademark of Ampco Metal Division, Ampco Pittsburgh Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?