• USA Home
  • Z179582 - Michel-Miller safety shield
EMAIL THIS PAGE TO A FRIEND
Z179582 Aldrich
Michel-Miller safety shield
size BB
Purchase
Description
General description
Ensures safe operation of M-M columns at pressures to 300 psi. Top thread of column is hung between two-piece aluminum cap; cap is then held by plastic shield. Clamp shield, not column. The 3 mm acrylic wall is not resistant to most solvents.
Price and Availability
All labs need water
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?