• USA Home
  • Z180335 - Tubing connector for large columns
EMAIL THIS PAGE TO A FRIEND
Z180335 Aldrich
Tubing connector for large columns
Insert, tube I.D. 1.5 mm
Purchase
Description
General description
TFE nut with 1/4-28 UNF 2B male thread connects small I.D. tubing to bottom drip adapter. KEL-F insert mechanically wedges tubing against fitting. For vacuum or pressure applications. Tubing-insertion tool required for assembly. Order nut, insert, and insertion tool separately.
Price and Availability
All labs need water
Non-Sterile Millex Syringe Filters
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?