Z181412 Aldrich
Perfektum® matched numbered hypodermic syringes
capacity 20 mL, needle-lock Luer
DOWNLOAD MSDS (PDF)
Properties
Related Categories Hypodermic Syringes, Labware, Perfektum Hypodermic Syringes, Syringes and Needles
feature   graduated 1.0 mL
mfr. no.   Popper 5137
capacity   20 mL
Description
General description
Perfektum® matched numbered syringes feature frosted individually lapped barrels and plungers which can withstand higher temperatures. These syringes are individually boxed and have matching serial numbers on mating barrels and plungers.
Z181455, Z181471, and z180378 include tip adapters to convert large male glass Luer to standard male metal Luer.
All are made from borosilicate glass. Metal parts are chrome-plated brass. Syringes have male fittings.
• Matched serial numbers on barrels and plungers for critical applications, not interchangeable
• Frosted, individually lapped barrels and plungers withstand some-what higher pressures
• Conform fully to U.S. Government specifications GG-S-921 Band GG-S-935
• Individually boxed
Other Notes
Because of the very close fit between the plunger and the barrel of the syringe, the slightest amount of dust or grit will scratch the smooth surfaces of the syringe. To prevent this both the plunger and barrel should be washed with a good detergent and dried with acetone prior to use. Store the clean dry syringe assembled, to prevent dust collecting on the plunger.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Customers Also Viewed
Perfektum<SUP>®</SUP> matched numbered hypodermic syringes
capacity 50 mL, needle-lock Luer
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?