• USA Home
  • Z181463 - Perfektum® matched numbered hypodermic syringes
EMAIL THIS PAGE TO A FRIEND
Z181463 Aldrich
Perfektum® matched numbered hypodermic syringes
capacity 50 mL, tip style, needle-lock Luer
  •  eCl@ss 32070500
Purchase
Properties
Related Categories Hypodermic Syringes, Labware, Perfektum Hypodermic Syringes, Syringes and Needles
feature   graduated 2.0 mL
  tip style needle-lock Luer
mfr. no.   Popper 5157
capacity   50 mL
Description
General description
Perfektum® matched numbered syringes feature frosted individually lapped barrels and plungers. These syringes are individually boxed and have matching serial numbers on mating barrels and plungers.
Z181455, Z181471 and Z180378 include tip adapters to convert large male glass Luer to standard male metal Luer.
• All are made from borosilicate glass
• Metal parts are chrome-plated brass
• Syringes have male fittings
• Matched serial numbers on barrels and plungers for critical applications, not interchangeable
• Frosted, individually lapped barrels and plungers withstand some-what higher pressures
• Individually boxed
Other Notes
Because of the very close fit between the plunger and the barrel of the syringe, the slightest amount of dust or grit will scratch the smooth surfaces of the syringe. Store the clean dry syringe assembled, to prevent dust collecting on the plunger.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?