• USA Home
  • Z181536 - Perfektum® eccentric-tip syringe
EMAIL THIS PAGE TO A FRIEND
Z181536 Aldrich
Perfektum® eccentric-tip syringe
capacity 10 mL, glass Luer
Purchase
Properties
Related Categories Hypodermic Syringes, Labware, Perfektum Hypodermic Syringes, Syringes and Needles
feature   graduated 0.2 mL
mfr. no.   Cadence Science 5126
capacity   10 mL
Description
General description
Eccentric tip syringes feature an offset nozzle that is on the edge of the syringe barrel. This allows an attached needle to be closer in line with the syringe walls so that the needle is nearly parallel with the injection surface. They are designed for use in procedures where the needle is to be introduced as closely parallel to injection area as possible. There are matched serial numbers on the plungers and barrels.
Other Notes
Because of the very close fit between the plunger and the barrel of the syringe, the slightest amount of dust or grit will scratch the smooth surfaces of the syringe. Store the clean dry syringe assembled, to prevent dust collecting on the plunger.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?