• USA Home
  • Z181609 - Perfektum® tuberculin syringe
EMAIL THIS PAGE TO A FRIEND
Z181609 Aldrich
Perfektum® tuberculin syringe
capacity 0.25 mL, glass Luer
  •  eCl@ss 32070500
Purchase
Properties
Related Categories Hypodermic Syringes, Labware, Perfektum Hypodermic Syringes, Syringes and Needles
feature   graduated 0.01 mL
mfr. no.   Cadence Science 5200
capacity   0.25 mL
Description
General description
Due to the very close fit between the plunger and the barrel of the syringe, the slightest amount of dust or grit will scratch the smooth surfaces of the syringe. To prevent this both the plunger and barrel should be washed with a good detergent and dried with acetone prior to use.
Store the clean dry syringe assembled, to prevent dust collecting on the plunger.
CAD5203 includes blue plunger to provide better contrast when dispensing opaque solutions.
Z181625 includes metal plunger.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?