• USA Home
  • Z181684 - Micro-Mate® interchangeable dual scale tuberculin syringes
EMAIL THIS PAGE TO A FRIEND
Z181684 Aldrich
Micro-Mate® interchangeable dual scale tuberculin syringes
capacity 2 mL (32 minims), needle-lock Luer
Purchase
Properties
Related Categories Dual-Scale Tuberculin Syringes, Labware, Microbiology Equipment, Syringes and Needles, Tuberculin Syringes,
feature   graduated 0.025 mL (0.5 minims)
mfr. no.   Popper 5207
capacity   2 mL (32 minims)
Description
General description
Because of the very close fit between the plunger and the barrel of the syringe, the slightest amount of dust or grit will scratch the smooth surfaces of the syringe.
Store the clean dry syringe assembled, to prevent dust collecting on the plunger.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?