• USA Home
  • Z181765 - Micro-Mate® pressure-control syringe
EMAIL THIS PAGE TO A FRIEND
Z181765 Aldrich
Micro-Mate® pressure-control syringe
capacity 5 mL, tip style, needle-lock Luer
Purchase
Properties
Related Categories Hypodermic Syringes, Labware, Micro-Mate Hypodermic Syringes, Syringes and Needles
feature   graduated 0.2 mL
  tip style needle-lock Luer
mfr. no.   Popper 5341
capacity   5 mL
Description
General description
Micro-Mate® pressure control glass syringes are designed with large, chrome-plated thumb and finger rings that afford positive grip and permit one-hand filling, and accurate delivery and pipetting. Syringes have needle-lock Luer tip and interchangeable parts. Pressure control syringes are particularly useful for injecting viscous fluids or in any application that requires one-hand filling.
Other Notes
Because of the very close fit between the plunger and the barrel of the syringe, the slightest amount of dust or grit will scratch the smooth surfaces of the syringe. To prevent this both the plunger and barrel should be washed with a good detergent and dried with acetone prior to use. Store the clean dry syringe assembled, to prevent dust collecting on the plunger.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?