• USA Home
  • Z181846 - Perfektum® jumbo syringe
EMAIL THIS PAGE TO A FRIEND
Z181846 Aldrich
Perfektum® jumbo syringe
capacity 150 mL, needle-lock Luer
Purchase
Properties
Related Categories Hypodermic Syringes, Labware, Perfektum Hypodermic Syringes, Syringes and Needles
feature   graduated 10 mL
mfr. no.   Cadence Science 5331
capacity   150 mL
Description
General description
Useful for dispensing, diluting, metering, mixing, pipetting, sampling, transferring, etc.
Other Notes
Because of the very close fit between the plunger and the barrel of the syringe, the slightest amount of dust or grit will scratch the smooth surfaces of the syringe. Store the clean dry syringe assembled, to prevent dust collecting on the plunger.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?