• USA Home
  • Z182133 - Perfektum® one-way spring-clip stopcock
EMAIL THIS PAGE TO A FRIEND
Z182133 Aldrich
Perfektum® one-way spring-clip stopcock
female Luer to male needle-lock Luer
Purchase
Properties
Related Categories Labware, Stopcocks, Syringe connectors, Syringes and Needles
feature   female Luer to male needle-lock Luer
mfr. no.   Popper 6067
Description
General description
Spring-clip design holds key in position under proper sealing pressure for optimum performance (need for screwdriver is eliminated) and facilitates disassembly. Precision-machined stopcock keys are individually lapped and mated to maintain pressure-test characteristics. Easily interfaced with assemblies of other stopcocks, manifolds, adapters, tubing needles, and glass syringes. Meet all U.S. Federal Specifications. Chrome-plated brass. Water tested at 60 psi.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?