• USA Home
  • Z182192 - Perfektum® one-way compression-nut stopcock
EMAIL THIS PAGE TO A FRIEND
Z182192 Aldrich
Perfektum® one-way compression-nut stopcock
Hose barb to hose barb
Purchase
Properties
Related Categories Labware, Stopcocks, Syringe connectors, Syringes and Needles
feature   For 1/16 to 3/32 in I.D. tubing
mfr. no.   Popper 6024
Description
General description
Water tested at 80 psi.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?