• USA Home
  • Z182265 - Adjustable needle stopper
EMAIL THIS PAGE TO A FRIEND
Z182265 Aldrich
Adjustable needle stopper
Fits 15 to 19 gauge needles
Purchase
Properties
Related Categories Labware, Luer Lock Syringe Connectors, Syringe connectors, Syringes and Needles
feature   Fits 15 to 19 gauge needles
mfr. no.   Popper 7573
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?