• USA Home
  • Z183458 - Oyster economy pH meter kit
EMAIL THIS PAGE TO A FRIEND
Z183458 Aldrich
Oyster economy pH meter kit
Purchase
Properties
Related Categories Extech, Labware, Manufacturer Browser, Portable Meters, pH Meters,
mfr. no.   Extech Instruments 120505
measuring range   0-14 pH unit pH, accuracy: 0.02 pH unit, resolution: 0.01 pH unit
Description
General description
LCD display in hinged Oyster cover adjusts for best viewing angle, shuts off power when closed. Runs on a 9V battery. Includes meter, polymer-body combination electrode, pH 4 and 7 buffer capsules and containers, neckstrap, and impact-resistant carrying case.
Temp. compensation: 0 to 60°C, manual
Legal Information
Oyster is a trademark of Extech Instruments Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?