• USA Home
  • Z183741 - Poly-net® protective netting

EMAIL THIS PAGE TO A FRIEND
Z183741 Aldrich

Poly-net® protective netting

red, diam. 5-8 in., Heavy-gauge

Purchase

Properties

Related Categories Films and Foils, Labware More...
diam.   5-8 in.
thickness   0.07 to 1.2 mm , approximate
color   red

Description

Application

Desiccators and vacuum flasks

General description

PE net sleeve stretches around glassware to prevent scratching,chipping or breaking. Netting captures flying glass if container shatters. Easily cut to any length. Color-coded for size identification.

Legal Information

Poly-net is a registered trademark of Norddeutsche Seekabelwerke GmbH & Co. KG

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?