• USA Home
  • Z184799 - Cochranes Fieldhouse Hiliwire bonds
EMAIL THIS PAGE TO A FRIEND
Z184799 Aldrich
Cochranes Fieldhouse Hiliwire bonds
Synonym: chemistry model sets, molecular model sets
Purchase
Description
General description
Coiled ribbon of 22-gauge vinyl-coated wire loops for making flexible connecting links with Orbit-size bonding tubes. Loops are cut from coil and inserted into tubes. Especially useful where non-standard bond angles are needed. Includes instruction booklet.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?