EMAIL THIS PAGE TO A FRIEND
Z185000 Aldrich
Prescored ampules
clear glass, volume 50 mL
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Ampules, Ampule, Ampules, Analytical/Chromatography,
material   clear glass
packaging   pkg of 144 ea
mfr. no.   Wheaton Scientific
volume   50 mL
Description
General description
Stems can be pull or tip sealed to effect a hermetically sealed container. Made from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements as defined by the Pharmacopeia of the United States. Pre-scored to eliminate filing.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?