• USA Home
  • Z187917 - Precisionware bottles
EMAIL THIS PAGE TO A FRIEND
Z187917 Aldrich
Precisionware bottles
Narrow-mouth, capacity 125 mL
Synonym: bottle hdpe, plastic bottle, plastic bottles
Purchase
Properties
Related Categories Bel-Art, Bel-Art Bottles and Bottle Accessories, Bottles and Caps, High-Density Polyethylene, Labware,
material   high-density polyethylene bottle
  polypropylene cap
packaging   pkg of 12 ea
capacity   125 mL
Description
General description
Bottle with leakproof and sturdy, heavy-duty wall. Manufactured to strict DOT-2E specifications. Molded pour lip for easy, no-spill pour. Suitable for shipping.
Legal Information
Precisionware is a trademark of Vectra Precisionware
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?