• USA Home
  • Z188042 - Filter-paper support discs
EMAIL THIS PAGE TO A FRIEND
Z188042 Aldrich
Filter-paper support discs
diam. 55 mm
Purchase
Properties
Related Categories Buchner Funnels, Funnels, Labware More...
diam.   55 mm
Description
General description
Made from high density polyethylene, these discs have excellent chemical resistance. They are useful for holding up filter paper. In this way the blocking of the holes is avoided and the filtering surface of the funnel is free. Will withstand temperatures up to 110°C.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?