EMAIL THIS PAGE TO A FRIEND
Z190314 Aldrich
Air-driven stirrer
X-Heavy usage, rpm, 8 ‑ 170
Synonym: air driven stirrers, air mixers, air stirrers
Purchase
Properties
Related Categories Labware, Overhead - SS Stirrers, Stirrers More...
feature   rpm 8 ‑ 170
availability   not available in EU
support rod diam. × L   9/16 in. × 10 in.
Description
General description
• Explosion-proof, stall-proof, does not overheat
• Exhaust muffler
• Brass needle valve with 1/4 in. NPTM outlet controls air intake
• Mount on standard clamp holders and vertical rods (order separately)
Includes: a screw collet and 3/8 in. diam. × 12 in. L SS stirrer shaft with propeller. A 3-jaw geared chuck may be ordered separately.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?