EMAIL THIS PAGE TO A FRIEND
Z190349 Aldrich
Powerhold clamp
swivel (with three adjustable knobs)
Purchase
Description
General description
With separate safety stop. Used for mounting rods 3/8 to 5/8 in. diameter. Made from high-strength machined aluminum rod with large knobs for gripping power.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?