• USA Home
  • Z192082 - Becton Dickinson polypropylene tuberculin syringes
EMAIL THIS PAGE TO A FRIEND
Z192082 Aldrich
Becton Dickinson polypropylene tuberculin syringes
Permanently affixed bevel point needle, capacity 0.5 mL
Purchase
Properties
Related Categories Becton Dickinson, Becton Dickinson Polypropylene Tuberculin Syringes, Labware, Manufacturer Browser, Polypropylene Tuberculin Syringes,
feature   graduated 0.01 mL
mfr. no.   Becton-Dickinson & Co.
capacity   0.5 mL
needle gauge × L   27 × 1/2 in.
Description
General description
Permanently affixed bevel point needle
Thinner and smaller syringe for easier control. Bold clear scale markings.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?