• USA Home
  • Z192449 - BD Precisionglide® syringe needles

EMAIL THIS PAGE TO A FRIEND
Z192449 Aldrich

BD Precisionglide® syringe needles

gauge 23, L 3/4 in.

Synonym: cannula

Purchase

Properties

Related Categories Becton Dickinson, Becton Dickinson Precisionglide Syringe Needles, Disposable syringes, Labware, Manufacturer Browser,
packaging   pkg of 100 ea
mfr. no.   Becton-Dickinson & Co. 305143
L   3/4 in.
gauge   23
color   turquoise

Description

General description

• Fits any Luer lock, slip tip, or eccentric tip syringe
• SS Microlance regular-bevel point
• Siliconized to reduce resistance
• PP Luer hub color coded to needle gauge
• Packaged as individual fiber-free, peel-apart packages in strips of five sterile needles
• A sheath protects point and cannula
• All packages carry lot control numbers to insure sterility
• NoKor needles have scalpel like tip

Legal Information

Precisionglide is a registered trademark of Becton-Dickinson & Co.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?