• USA Home
  • Z192589 - STACO variable transformer
EMAIL THIS PAGE TO A FRIEND
Z192589 Aldrich
STACO variable transformer
Ventilated steel-case
Purchase
Properties
Related Categories Heating, Heating Controllers, Labware More...
availability   not available in EU
output   10 A
Description
General description
Suitable for controlling electric heating elements, lights, electric motor speed, or power. Output voltage controlled by a large slip-resistant knob with the output increasing linearly as the knob is turned clockwise. All models include a three-conductor grounded line cord, matching receptacle, switch, pilot lamp, and fuse.
Transformers include a lighted rocker switch that is either off or 0-140 Volts.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?