• USA Home
  • Z192597 - STACO variable transformer
EMAIL THIS PAGE TO A FRIEND
Z192597 Aldrich
STACO variable transformer
Ventilated steel-case with 0-150V volt meter
Purchase
Properties
Related Categories Heating, Heating Controllers, Labware More...
availability   not available in EU
Description
General description
Suitable for controlling electric heating elements, lights, electric motor speed, or power. Output voltage controlled by a large slip-resistant knob with the output increasing linearly as the knob is turned clockwise. All models include a three-conductor grounded line cord, matching receptacle, switch, pilot lamp, and fuse.
Transformers include a lighted rocker switch that is either off or 0-140 Volts.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?