EMAIL THIS PAGE TO A FRIEND
Z192783 Aldrich
Microtip probe
tip diam. 2 mm, sample volume 0.15-5 mL
Purchase
Properties
mfr. no.   Sonics & Materials 630-0423
sample volume   0.15-5 mL
tip diam.   2 mm
Description
General description
For processing small samples at extremely high-intensity. Probes can reach down into Eppendorf tubes, culture tubes, scintillation vials, etc. Made of titanium, autoclavable.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?