EMAIL THIS PAGE TO A FRIEND
Z192813 Aldrich
Micro cup horn
I.D. 1 in. (25 mm)
Purchase
Properties
Related Categories Branson processors, Labware, Manufacturer Browser, Sonics & Materials, Sonics & Materials Horns and Horn Tips,
mfr. no.   Sonics & Materials 630-0447
I.D.   1 in. (25 mm)
Description
General description
For processing liquids in absolute isolation without probe intrusion. Useful for work with pathogenic, radioactive, or biohazardous samples. The water-filled micro cup horn screws into the converter. Vibrations transmitted through the water in the cup induce cavitation inside the test tube immersed in the water.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?