• USA Home
  • Z195847 - Ansell Edmont shoulder-length neoprene gloves
EMAIL THIS PAGE TO A FRIEND
Z195847 Aldrich
Ansell Edmont shoulder-length neoprene gloves
Purchase
Properties
material   black neoprene
size   10
mfr. no.   Ansell Edmont
L   31 in.
Description
General description
Gauntlet style gloves made of neoprene bonded to a soft, absorbent interior cotton shell. Curved fingers. One size fits all.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z170070 Economical neoprene dry-box gloves, size 10, port size 8 in.
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?