• USA Home
  • Z197696 - Justrite® safety funnel
EMAIL THIS PAGE TO A FRIEND
Z197696 Aldrich
Justrite® safety funnel
shaft L 32 in., perforated
Purchase
Properties
Related Categories Justrite, Justrite Funnels, Labware, Manufacturer Browser, Safety Equipment,
mfr. no.   Justrite 8205
shaft L   32 in.
Description
General description
Large 10 3/4 in. mouth. Self-closing drum funnels provide quick dumping of flammable solvents while minimizing dangerous spills. The flame-arresting brass shaft and hinged cover with fusible link reduce spread of hazardous vapors. If ignition occurs in the open position, the fusible link melts at 135°F causing the lid to snap shut. 18 gauge terneplated.
For solvent disposal applications.
Legal Information
Justrite is a registered trademark of Justrite Manufacturing Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?