• USA Home
  • Z199834 - Oberon chemically-resistant safety goggles
EMAIL THIS PAGE TO A FRIEND
Z199834 Aldrich
Oberon chemically-resistant safety goggles
Indirect vent/antifog lens
Purchase
Properties
Related Categories Eye Protection Equipment, Goggles, Labware, Safety Equipment
material   polycarbonate lens
  clear vinyl frame
availability   not available in EU
lens thickness   0.050 in.
Description
General description
Resist mild acids, caustics, aromatics, hydrocarbons, and methylene chloride (excludes hydrofluoric acid). Coated with chemically resistant Silicote-6. Meets ANSI Z87.1-1989 requirements.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?