• USA Home
  • Z201065 - Spilfyter® hazmat spill control, polypropylene
EMAIL THIS PAGE TO A FRIEND
Z201065 Aldrich
Spilfyter® hazmat spill control, polypropylene
Sheet, W × L 24 in. × 24 in.
Purchase
Properties
Related Categories Aldrich, Aldrich Safety Equipment, Labware, Manufacturer Browser, Pillows and Socks,
packaging   pkg of 10 ea
  pkg of 50 ea
W × L   24 in. × 24 in.
Description
General description
Spilfyter® products are made from a specially formulated, lint-free, 100% PP fabric that is virtually inert and will absorb any liquid with which they come in contact. They are able to collect up to 10 times their own weight in waste material (depending upon its viscosity) in 30s. Saturated Spilfyter® products can be landfilled, neutralized, or incinerated in accordance with federal, state and local regulations.
Legal Information
Spilfyter is a registered trademark of National Packaging Services Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z158054 Spilfyter® hazmat spill control, polypropylene, Pillow, capacity 4 L
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?