EMAIL THIS PAGE TO A FRIEND
Z201901 Aldrich
Remco scraper
L × W 9 1/4 in. × 4 5/16 in.
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Remco, Spatulas and Scoops
L × W   9 1/4 in. × 4 5/16 in.
Description
General description
One-piece, corrosion-resistant PP. Non-abrasive, will not scratch SS. Ideal for countertops, vials, etc. Can be sterilized.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?