• USA Home
  • Z202525 - Aldrich® solvent-still-head apparatus
EMAIL THIS PAGE TO A FRIEND
Z202525 Aldrich
Aldrich® solvent-still-head apparatus
flask capacity 250 mL
Purchase
Description
General description
24/40 joints. Includes still head, column with Raschig rings, Friedrichs condenser, one-neck round-bottom flask, stopper, and nitrogen-inlet adapter. Designed by Paul F. Hudrlik. May be used for continuous distillation.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?