• USA Home
  • Z203467 - Framework Molecular Model (FMM) set
EMAIL THIS PAGE TO A FRIEND
Z203467 Aldrich
Framework Molecular Model (FMM) set
includes instruction manual and plastic storage box
Purchase
Properties
Related Categories Framework models, Labware, Molecular Models, Molecular Models and Educational Aids
mfr. no.   Pearson Education
Description
General description
FMM sets utilize two basic construction units to represent all atoms in the periodic table:
• Valence clusters−includes tetrahedron, trigonal bipyramid, and octahedron. Clusters are made of metal and are color-coded: silver, brass, and copper.
• Bond tubes−color-coded plastic tubes fit snugly onto prongs of valence clusters.
• Note: Includes instruction manual and plastic storage box.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?