• USA Home
  • Z205079 - Eagle lab safety can
EMAIL THIS PAGE TO A FRIEND
Z205079 Aldrich
Eagle lab safety can
capacity 1 gal
Purchase
Properties
Related Categories Eagle, Labware, Manufacturer Browser, Metal Safety Containers, Safety Containers,
material   terneplated steel
mfr. no.   Eagle
availability   not available in EU
H × W   13 in. × 6 in.
capacity   1 gal
Description
General description
FM approved. Meets OSHA requirements. Electrically welded construction with rolled edge pour spout, flame arrester, and PTFE gasket. Oval shape conserves space.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?