• USA Home
  • Z205109 - Laboratory shelf can
EMAIL THIS PAGE TO A FRIEND
Z205109 Aldrich
Laboratory shelf can
capacity 1 gal
Purchase
Properties
Related Categories Justrite, Justrite Safety, Labware, Manufacturer Browser, Metal Safety Containers,
material   terneplated steel
mfr. no.   Justrite 10307
availability   not available in EU
H × diam.   4 5/8 in. × 6 3/4 in.
capacity   1 gal
Description
General description
Safety can with self-closing brass faucet for controlled dispensing of hazardous liquids. Faucet is mounted two inches above bottom. FM approved.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?