• USA Home
  • Z205117 - Justrite® nonmetallic lab-shelf can
EMAIL THIS PAGE TO A FRIEND
Z205117 Aldrich
Justrite® nonmetallic lab-shelf can
Purchase
Properties
Related Categories Justrite, Justrite Safety, Labware, Manufacturer Browser, Plastic Safety Containers,
material   red high-density polyethylene
mfr. no.   Justrite 14590
H × O.D.   16 in. × 12 3/4 in.
capacity   5 gal
Description
General description
Safety can with self-closing faucet for controlled dispensing of hazardous liquids. Self-closing pressure-relief cap has built-in vacuum breaker and double-mesh flame arrester. Round can has a brass faucet mounted 2 in. above bottom.
Legal Information
Justrite is a registered trademark of Justrite Manufacturing Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?