• USA Home
  • Z208205 - Rubbermaid® shelf for heavy-duty lab carts

EMAIL THIS PAGE TO A FRIEND
Z208205 Aldrich

Rubbermaid® shelf for heavy-duty lab carts

middle shelf for C, beige

Synonym: cart shelf

Purchase

Properties

Related Categories Accessories, Carboys & Cans, Labware, Manufacturer Browser, Other Racks,
mfr. no.   Rubbermaid
color   beige

Description

Legal Information

Rubbermaid is a registered trademark of Rubbermaid Commercial Products, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?