• USA Home
  • Z208973 - Fiesers’ Reagents for Organic Synthesis
EMAIL THIS PAGE TO A FRIEND
Z208973 Aldrich
Fiesers’ Reagents for Organic Synthesis
Volume 15
  •  ISBN-10 0471521132
  •  ISBN-13 978-047152113-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   M. Fieser, 1990, 432 pp., hard cover
mfr. no.   Wiley
Description
General description
The popular Fieser and Fieser series on reagents for organic synthesis provides concise descriptions, good structural formulas and selected examples of applications--providing references to new reagents as well as to reagents included in previous volumes. Thousands of entries abstract the most important information on commonly used and new reagents, including preparation, uses, sources of supply, critical comments, references and more. The reagents are considered in alphabetical order by common usage names
Thousands of entries abstract the most important information on commonly used reagents. Every reagent discussed includes the preparation, uses, sources of supply, critical comments, references, and more.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?