• USA Home
  • Z210102 - Digi-Block® digital block heater
EMAIL THIS PAGE TO A FRIEND
Z210102 Aldrich
Digi-Block® digital block heater
AC input 210 - 230 AC
Purchase
Properties
feature   capacity 4 blocks
  temp. range≤ 200 °C (by 0.1 °C)
mfr. no.   Barnstead Thermolyne
H × L × W   12 cm × 39 cm × 17 cm
weight   4.5 kg
output   300 W
Description
General description
Provides safe, dry heating for clinical and general chemistry applications. Interchangeable heating blocks accommodate test tubes, microcentrifuge tubes, and cuvettes. Order blocks separately.
• Control panel displays "actual" and "set" temperatures
• Non-volatile memory retains temperature settings
• Aluminum and SS housing will not melt or rust
• Massive aluminum heating plate assures even heating
• Platinum RTD probe fits into heating blocks next to samples for precise temperature control
Legal Information
Digi-Block is a registered trademark of Laboratory Devices, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?