• USA Home
  • Z210153 - Digi-Block® heating block
EMAIL THIS PAGE TO A FRIEND
Z210153 Aldrich
Digi-Block® heating block
For test tubes, no. of holes, 24, for tube diam., 12 ‑ 13 mm
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Block Heaters, Barnstead Thermolyne Heating Equipment, Block Heaters, Digi-Block blocks for block heaters,
feature   for tube diam. 12 ‑ 13 mm
  no. of holes 24
mfr. no.   Barnstead International 5037
Description
General description
Precision machined, solid aluminum heating blocks for use with DIGI-BLOCK heater. Interchangeable.
Legal Information
Digi-Block is a registered trademark of Laboratory Devices, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?