• USA Home
  • Z210188 - Digi-Block® heating block
EMAIL THIS PAGE TO A FRIEND
Z210188 Aldrich
Digi-Block® heating block
For test tubes, no. of holes, 6, for tube diam., 20 mm
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Block Heaters, Barnstead Thermolyne Heating Equipment, Block Heaters, Digi-Block blocks for block heaters,
feature   for tube diam. 20 mm
  no. of holes 6
mfr. no.   Barnstead International 5038
Description
General description
Precision machined, solid aluminum heating blocks for use with DIGI-BLOCK heater. Interchangeable.
Legal Information
Digi-Block is a registered trademark of Laboratory Devices, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?