• USA Home
  • Z210560 - Pro-Pak® distillation packing
EMAIL THIS PAGE TO A FRIEND
Z210560 Aldrich
Pro-Pak® distillation packing
0.24 in.2, Monel
Purchase
Description
General description
Efficient, self-wetting, protruded metal dump-packing for distillation columns of 0.75 to 12 in. diameter. Maximum reflux rate of approximately 3.4 L/h. Free space for 0.16 in.2 size is 94% and for 0.24 in.2 size is 96%. Each package has a packing volume of approx. 1 L. Packed heights up to 20 ft possible without distortion.
Legal Information
Pro-Pak is a registered trademark of Cannon Instruments Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?