• USA Home
  • Z210862 - 3M disposable respirator
EMAIL THIS PAGE TO A FRIEND
Z210862 Aldrich
3M disposable respirator
dusts and mists, prefilter retainer
Purchase
Properties
Related Categories 3M, Disposable Respirators, Disposable respirator, Labware, Manufacturer Browser,
packaging   pkg of 10 ea
mfr. no.   3M 5000
Description
General description
Pre-assembled disposable respirators. Low-profile design provides improved comfort and less vision interference. Approved for: Organic vapor/acid gas and dusts/mists having a TWA not less than 0.05 mg/m3 or 2 mppcf.
NIOSH approved.
Protection factor: Up to 10 × PEL.
Legal Information
3M is a trademark of 3M Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?