• USA Home
  • Z210870 - 3M disposable respirator
EMAIL THIS PAGE TO A FRIEND
Z210870 Aldrich
3M disposable respirator
dusts and mists, prefilter retainer
Purchase
Properties
Related Categories 3M, Disposable Respirators, Disposable respirator, Labware, Manufacturer Browser,
packaging   pkg of 2 ea
mfr. no.   3M 5000
Description
General description
Pre-assembled disposable respirators. Low-profile design provides improved comfort and less vision interference. Approved for: Organic vapor/acid gas and dusts/mists having a TWA not less than 0.05 mg/m3 or 2 mppcf.
NIOSH approved.
Protection factor: Up to 10 × PEL.
Legal Information
3M is a trademark of 3M Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?