• USA Home
  • Z214000 - Aldrich® Vapor-Phase FT-IR Databases
EMAIL THIS PAGE TO A FRIEND
Z214000 Aldrich
Aldrich® Vapor-Phase FT-IR Databases
w/FT-IR book, hard cover/CD-ROM
Purchase
Properties
Related Categories Computer Software, Labware, Spectral Libraries More...
publication info   , hard cover/CD-ROM
Description
General description
• High resolution vapor-phase FT-IR spectral library for use with Spectral I.D®, GRAMS®, and other spectral search software. The library contains spectra at 4 cm-1 resolution by 16-bit ordinate precision, which are suitable for doing searches using spectral subtraction
• More than 5,000 FT-IR spectra of organics produced using high resolution gas chromatography
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
GRAMS is a registered trademark of Thermo Electron Corp.
Spectral ID is a registered trademark of Galactic Industries Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?